missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Piperidineethanol, TRC

Product Code. 30411063
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411063 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411063 Supplier Toronto Research Chemicals Supplier No. TRCP481955100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-Piperidineethanol
CAS 103639-57-2
Molecular Formula C7H15NO
Quantity 100 mg
Synonym (2S)-2-Piperidineethanol, (-)-2-Piperidineethanol, (S)-2-(2-Hydroxyethyl)piperidine
InChI Formula InChI=1S/C7H15NO/c9-6-4-7-3-1-2-5-8-7/h7-9H,1-6H2/t7-/m0/s1
SMILES OCC[C@@H]1CCCCN1
IUPAC Name 2-[(2S)-piperidin-2-yl]ethanol
Molecular Weight (g/mol) 129.2
Formula Weight 129.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.