missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Phenyl-1-propanol, 98+%

Product Code. 11378865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378865 1 g 1g
11388865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378865 Supplier Thermo Scientific Alfa Aesar Supplier No. L13988.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37778-99-7 | C9H12O | 136.194 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37778-99-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00145249
InChI Key RNDNSYIPLPAXAZ-MRVPVSSYSA-N
Synonym s-2-phenylpropan-1-ol, s---2-phenyl-1-propanol, --2-phenylpropanol, s-2-phenyl-1-propanol, 2s-2-phenylpropan-1-ol, unii-w7q89429zo, 2 s-phenyl-1-propanol, --2-phenyl-1-propanol, --beta-methylbenzeneethanol
PubChem CID 447661
IUPAC Name (2S)-2-phenylpropan-1-ol
SMILES CC(CO)C1=CC=CC=C1

Specifications

Density 0.975
Boiling Point 220°C
Flash Point 108°C (226°F)
Refractive Index 1.526
Quantity 1 g
Optical Rotation −18° (Neat)
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Phenyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.