missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Pentanol, 97%

Product Code. 11357336
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357336 250 mg 250mg
11347336 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357336 Supplier Thermo Scientific Alfa Aesar Supplier No. L09314.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26184-62-3 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26184-62-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00065952
InChI Key JYVLIDXNZAXMDK-YFKPBYRVSA-N
Synonym s-+-2-pentanol, s-2-pentanol, 2s-pentan-2-ol, 2-pentanol, 2s, s---2-pentanol, 2s-2-pentanol, s +-2-pentanol
PubChem CID 2724896
IUPAC Name (2S)-pentan-2-ol
SMILES CCCC(C)O

Specifications

Density 0.81
Boiling Point 118°C to 119°C
Flash Point 33°C (91°F)
Refractive Index 1.406
Quantity 250 mg
UN Number UN1105
Beilstein 1718820
Optical Rotation +14° (Neat)
Formula Weight 88.15
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Pentanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.