missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Octyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 15469119
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15469119

Brand: Thermo Scientific Alfa Aesar L19985.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737761-98-7
Density 0.907
Boiling Point 95°C to 96°C (4 mmHg)
Molecular Formula C9H17NS
Refractive Index 1.484
MDL Number MFCD04039337
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym s-+-2-octyl isothiocyanate, 2s-2-isothiocyanatooctane
InChI Key RXAAZXUXIQEORL-VIFPVBQESA-N
SMILES CCCCCCC(C)N=C=S
IUPAC Name (2S)-2-isothiocyanatooctane
Molecular Weight (g/mol) 171.302
PubChem CID 7016583
Formula Weight 171.31
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Octyl isothiocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.