missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Octanol, 99%

Product Code. 11398795
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398795 1 g 1g
11308805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398795 Supplier Thermo Scientific Alfa Aesar Supplier No. L12425.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6169-06-8 | C8H18O | 130.23 g/mol

(S)-(+)-2-Octanol is used as chiral compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Octanol is used as chiral compounds.

Solubility
Soluble in water. (1.12 g/L) at 25°C

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6169-06-8
MDL Number MFCD00064283
Synonym s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s
PubChem CID 2723888
ChEBI CHEBI:37870

Specifications

Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Molecular Formula C8H18O
Refractive Index 1.426
Quantity 1 g
Beilstein 1719323
Merck Index 14,6752
Solubility Information Soluble in water. (1.12g/L) at 25°C
InChI Key SJWFXCIHNDVPSH-UHFFFAOYNA-N
Optical Rotation +10° (c=1 in Ethanol)
SMILES CCCCCCC(C)O
IUPAC Name (2S)-octan-2-ol
Molecular Weight (g/mol) 130.23
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Octanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.