missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Nonyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 15449119
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15449119 Supplier Thermo Scientific Alfa Aesar Supplier No. L19983.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737000-86-1
Density 0.904
Molecular Formula C10H19NS
Refractive Index 1.4835
MDL Number MFCD04039335
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym s-+-2-nonyl isothiocyanate, 2s-2-isothiocyanatononane
InChI Key MQPOQQFZEYBCRM-JTQLQIEISA-N
SMILES CCCCCCCC(C)N=C=S
IUPAC Name (2S)-2-isothiocyanatononane
Molecular Weight (g/mol) 185.329
PubChem CID 53485810
Formula Weight 185.33
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Nonyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.