missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Methylbutyryl-CoA Sodium Salt, TRC

Product Code. 30476087
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476087 5 mg 5mg
30465167 1 mg 1mg
30470879 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476087 Supplier Toronto Research Chemicals Supplier No. TRCM1012905MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-Methylbutyryl-CoA Sodium Salt
Molecular Formula C26H44N7O17P3S . xNa
Quantity 5 mg
Synonym S-(2-(3-((2R)-4-(((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-hydroxy-3-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)oxy)-2-hydroxy-3,3-dimethylbutanamido)propanamido)ethyl) (2S)-2-Methylbutanethioate Sodium Salt
InChI Formula InChI=1S/C26H44N7O17P3S.Na/c1-5-14(2)25(38)54-9-8-28-16(34)6-7-29-23(37)20(36)26(3,4)11-47-53(44,45)50-52(42,43)46-10-15-19(49-51(39,40)41)18(35)24(48-15)33-13-32-17-21(27)30-12-31-22(17)33;/h12-15,18-20,24,35-36H,5-11H2,1-4H3,(H,28,34)(H,29,37)(H,42,43)(H,44,45)(H2,27,30,31)(H2,39,40,41);/q;+1/p-1/t14-,15+,18+,19+,20-,24+;/m0./s1
SMILES O[C@@H]([C@H](N1C2=NC=NC(N)=C2N=C1)O[C@@H]3COP(OP(OCC(C)([C@H](C(NCCC(NCCSC([C@H](CC)C)=O)=O)=O)O)C)(O)=O)(O)=O)[C@@H]3OP(O)(O[Na])=O
IUPAC Name sodium (2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-hydroxy-2-(((hydroxy((hydroxy((R)-3-hydroxy-2,2-dimethyl-4-((3-((2-(((S)-2-methylbutanoyl)thio)ethyl)amino)-3-oxopropyl)amino)-4-oxobutoxy)phosphoryl)oxy)phosphoryl)oxy)methyl)tetrahydrofuran-3-yl hydrogen phosphate
Molecular Weight (g/mol) 873.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.