missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylbutyric acid, 98%

Product Code. 10157793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157793 25 g Glass bottle 25g
10308410 1 g Glass bottle 1g
10687382 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10157793 Supplier Thermo Scientific Acros Supplier No. 224190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1730-91-2 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1730-91-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00063166
InChI Key WLAMNBDJUVNPJU-BYPYZUCNSA-N
Synonym s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
PubChem CID 448893
ChEBI CHEBI:38655
IUPAC Name (2S)-2-methylbutanoic acid
SMILES CCC(C)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Boiling Point 78.0°C (15.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COOH
Refractive Index 1.4041 to 1.4061
Quantity 25 g
Beilstein 02, 304
Specific Gravity 0.938
Specific Rotation Condition +19° (20°C c=neat)
Specific Rotation + 19.00
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.