missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methylbutylamine, 98+%

Product Code. 11318945
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318945 250 mg 250mg
11308945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318945 Supplier Thermo Scientific Alfa Aesar Supplier No. L14063.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34985-37-0 | C5H13N | 87.166 g/mol

(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34985-37-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00064430
InChI Key VJROPLWGFCORRM-YFKPBYRVSA-N
Synonym s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride
PubChem CID 2724272
IUPAC Name (2S)-2-methylbutan-1-amine
SMILES CCC(C)CN

Specifications

Boiling Point 96°C to 97°C
Flash Point 3°C (37°F)
Quantity 250 mg
UN Number UN2733
Beilstein 1718807
Optical Rotation −5.9° (Neat)
Formula Weight 87.16
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Methylbutylamine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-2-Methylbutylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.