missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methoxypropionic Acid, TRC

Product Code. 30482989
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482989 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482989 Supplier Toronto Research Chemicals Supplier No. TRCM26695525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(-)-2-Methoxypropionic Acid
CAS 23953-00-6
Molecular Formula C4 H8 O3
Quantity 25 g
Synonym Propanoic acid','2-methoxy-','(2S)-, Propanoic acid','2-methoxy-','(S)-, Propionic acid','2-methoxy-','(S)-(-)- (8CI), (2S)-2-Methoxypropanoic acid, (S)-(-)-2-Methoxypropionic acid, (S)-2-Methoxypropanoic acid
InChI Formula InChI=1S/C4H8O3/c1-3(7-2)4(5)6/h3H,1-2H3,(H,5,6)/t3-/m0/s1
SMILES CO[C@@H](C)C(=O)O
IUPAC Name (2S)-2-methoxypropanoic acid
Molecular Weight (g/mol) 104.1
Formula Weight 104.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.