missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methoxypropionic acid, 98%

Product Code. 10776242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10776242 500 mg Glass bottle 500mg
10515301 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10776242 Supplier Thermo Scientific Acros Supplier No. 331725000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23953-00-6 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23953-00-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD01632589
InChI Key ICPWFHKNYYRBSZ-VKHMYHEASA-N
Synonym s-2-methoxypropanoic acid, s---2-methoxypropionic acid, 2s-2-methoxypropanoic acid, propanoic acid, 2-methoxy-, 2s, unii-74k293ocx6, 2-methoxypropanoic acid, s, propanoic acid,2-methoxy-, 2s, propanoic acid, 2-methoxy-, s, propionic acid, 2-methoxy-, s--, s-2-methoxypropionic acid
PubChem CID 5702566
IUPAC Name (2S)-2-methoxypropanoic acid
SMILES CC(C(=O)O)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 99.0°C (20.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)COOH
Refractive Index 1.4132 to 1.4152
Quantity 500 mg
Beilstein 03, III, 444
Specific Rotation Condition −75.00 (20.00°C c=neat)
Specific Rotation − 75.00
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Methoxypropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.