missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methoxypropanol, 97%

Product Code. 10328653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
2.5 mL
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1mL
2.5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10328653 10 mL Glass bottle 10mL
10726622 1 mL Glass bottle 1mL
10244751 2.5 mL Glass bottle 2.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10328653 Supplier Thermo Scientific Supplier No. 331850100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116422-39-0 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116422-39-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD01632593
InChI Key YTTFFPATQICAQN-BYPYZUCNSA-N
Synonym 2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
PubChem CID 642402
IUPAC Name (2S)-2-methoxypropan-1-ol
SMILES CC(CO)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH2OH
Refractive Index 1.4060 to 1.4080
Quantity 10 mL
Specific Gravity 0.938
Specific Rotation Condition + 39.20 (20.00°C c=1.66,CHCl3)
Specific Rotation + 39.20
Formula Weight 90.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methoxypropanol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.