missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee

Product Code. 10111242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10111242 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10111242 Supplier Thermo Scientific Acros Supplier No. 362110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 208113-95-5 | C8H14O4 | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 208113-95-5
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O4
MDL Number MFCD01091031
Quantity 1 g
Synonym s-2-isopropylsuccinic acid-1-methyl ester, s-3-methoxycarbonyl-4-methylpentanoic acid, s-2-isopropylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-4-methylpentanoic acid, butanedioic acid, 2-1-methylethyl-, 1-methyl ester, 2s, s-2-isopropylsuccinic acid-1-methylester, 2-s-isopropylsuccinic acid 1-methyl ester, s-+-2-isopropylsuccinic acid 1-methyl ester
InChI Key VHOGSWBAPDPWTG-LURJTMIESA-N
SMILES CC(C)C(CC(=O)O)C(=O)OC
IUPAC Name (3S)-3-methoxycarbonyl-4-methylpentanoic acid
Molecular Weight (g/mol) 174.19
PubChem CID 2734514
Formula Weight 174.19
Percent Purity 95%,98% ee
Physical Form Viscous Liquid
Chemical Name or Material (S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.