missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific™

Product Code. 10324742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10324742 1 g Glass Bottle 1g
10095321 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324742 Supplier Thermo Scientific Acros Supplier No. 362100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213270-36-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD01091033
InChI Key AJNNFMRCMKGYHA-ZETCQYMHSA-N
Synonym s-3-methoxycarbonyl-5-methylhexanoic acid, s---2-isobutylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-5-methylhexanoic acid, s-2-isobutylsuccinic acid 1-methyl ester, s-2-2-methylpropyl butanedioic acid 1-methyl ester, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
PubChem CID 2734513
IUPAC Name (3S)-3-methoxycarbonyl-5-methylhexanoic acid
SMILES CC(C)CC(CC(=O)O)C(=O)OC

Specifications

Packaging Glass Bottle
Quantity 1 g
Formula Weight 188.22
Percent Purity 95%,(98% ee)
Chemical Name or Material (S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
compliance-icons
Product Identifier
  • (S)-2-Isobutylsuccinic acid-1-methyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.