missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Hydroxyethyl)glutathione, TRC

Product Code. 30394445
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394445 25 mg -
30395202 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394445 Supplier Toronto Research Chemicals Supplier No. TRCH94215625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28747-20-8
Molecular Formula C12H21N3O7S
Molecular Weight (g/mol) 351.38
Synonym L-gamma-Glutamyl-S-(2-hydroxyethyl)-L-cysteinylglycine, L-N-[1-[(Carboxymethyl)carbamoyl]-2-[(2-hydroxyethyl)thio]ethyl]glutamine, N-[N-L-gamma-Glutamyl-S-(2-hydroxyethyl)-L-cysteinyl]glycine, S-(beta-Hydroxyethyl)glutathione
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-(2-hydroxyethylsulfanyl)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
SMILES N[C@@H](CCC(=O)N[C@@H](CSCCO)C(=O)NCC(=O)O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H21N3O7S/c13-7(12(21)22)1-2-9(17)15-8(6-23-4-3-16)11(20)14-5-10(18)19/h7-8,16H,1-6,13H2,(H,14,20)(H,15,17)(H,18,19)(H,21,22)/t7-,8-/m0/s1
Formula Weight 351.11
Chemical Name or Material S-(2-Hydroxyethyl)glutathione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.