missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxy-3-buten-1-yl p-tosylate, 99%, Thermo Scientific™

Product Code. 10475461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10475461 1 g Glass bottle 1g
11378601 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10475461 Supplier Thermo Scientific Acros Supplier No. 411970010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 133095-74-6
Molecular Formula C11H14O4S
Molecular Weight (g/mol) 242.29
MDL Number MFCD00145259
InChI Key YQSCDBZHHLIPOI-JTQLQIEISA-N
Synonym s-3-butene-1,2-diol-1-p-toluenesulfonate, s-2-hydroxy-3-butenyl tosylate, 3-butene-1,2-diol, 1-4-methylbenzenesulfonate , 2s, 2s-1-4-methylbenzenesulfonyl oxy but-3-en-2-ol, pubchem6792, s-3-butene-1,2-diol 1-tosylate, 2s-2-hydroxybut-3-enyl 4-methylbenzenesulfonate, s-4-4-methylphenyl sulfonyl oxy-1-butene-3-ol, s-toluene-4-sulfonic acid 2-hydroxy-but-3-enyl ester, 3-butene-1,2-diol,1-4-methylbenzenesulfonate , 2s
PubChem CID 7015761
IUPAC Name [(2S)-2-hydroxybut-3-enyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H](O)C=C

Specifications

Melting Point 57.0°C to 63.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition − 78.50 (20.00°C c=1.13 MeOH)
Specific Rotation − 78.50
Formula Weight 242.3
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-2-Hydroxy-3-buten-1-yl p-tosylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.