missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Heptyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 15409129
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409129 Supplier Thermo Scientific Alfa Aesar Supplier No. L20065.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737000-94-1
Density 0.91
Molecular Formula C8H15NS
Refractive Index 1.486
MDL Number MFCD05664052
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym s-+-2-heptyl isothiocyanate, s-2-isothiocyanatoheptane, 2s-2-isothiocyanatoheptane, s-+-2-heptylisothiocyanate
InChI Key UPTLUDLHIIYPNC-QMMMGPOBSA-N
SMILES CCCCCC(C)N=C=S
IUPAC Name (2S)-2-isothiocyanatoheptane
Molecular Weight (g/mol) 157.275
PubChem CID 7016582
Formula Weight 157.28
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Heptyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.