missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Fmoc-amino)butyric acid, 98%, Thermo Scientific™

Product Code. 15443398
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15443398

Brand: Thermo Scientific Alfa Aesar H56852.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135112-27-5
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
MDL Number MFCD00080268
InChI Key XQIRYUNKLVPVRR-KRWDZBQOSA-N
Synonym fmoc-abu-oh, fmoc-2-abu-oh, fmoc-l-2-aminobutyric acid, fomc-abu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-2-aminobutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, s, l-2-fmoc-amino butyric acid
PubChem CID 853016
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 250 mg
Beilstein 7657232
Optical Rotation −22° (c=1 in DMF)
Formula Weight 325.36
Percent Purity 98%
Chemical Name or Material (S)-2-(Fmoc-amino)butyric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.