missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2-Chloropropionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-2-Chloropropionic Acid |
| CAS | 29617-66-1 |
| Molecular Formula | C3 H5 Cl O2 |
| Quantity | 1 g |
| Synonym | Propanoic acid, 2-chloro-, (2S)- (9CI, ACI), (2S)-2-Chloropropanoic acid (ACI), Propanoic acid, 2-chloro-, (S)- (ZCI), Propionic acid, 2-chloro-, L- (8CI), (-)-2-Chloropropanoic acid, (-)-2-Chloropropionic acid, (-)-α-Chloropropanoic acid, (2S)-2-Chloropropanoic acid, (2S)-2-Chloropropionic acid, (2S)-Chloropropanoic acid, (S)-2-Chloropropanoic acid, (S)-2-Chloropropionic acid, (S)-α-Chloropropionic acid, 2(S)-Chloropropionic acid, 2-Chloro-L-propionic acid, L-2-Chloropropanoic acid, L-2-Chloropropionic acid, S-(-)-2-Chloropropionic acid, α-L-Chloropropionic acid |
| InChI Formula | InChI=1S/C3H5ClO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m0/s1 |
| SMILES | C[C@H](Cl)C(=O)O |
| IUPAC Name | (2S)-2-chloropropanoic acid |
| Molecular Weight (g/mol) | 108.52 |
| Formula Weight | 107.9978 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?