missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic Acid, TRC

Product Code. 30362033
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362033 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362033 Supplier Toronto Research Chemicals Supplier No. TRCC3797002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(-)-2-Chloropropionic Acid
CAS 29617-66-1
Molecular Formula C3 H5 Cl O2
Quantity 2 g
Synonym Propanoic acid, 2-chloro-, (2S)- (9CI, ACI), (2S)-2-Chloropropanoic acid (ACI), Propanoic acid, 2-chloro-, (S)- (ZCI), Propionic acid, 2-chloro-, L- (8CI), (-)-2-Chloropropanoic acid, (-)-2-Chloropropionic acid, (-)-α-Chloropropanoic acid, (2S)-2-Chloropropanoic acid, (2S)-2-Chloropropionic acid, (2S)-Chloropropanoic acid, (S)-2-Chloropropanoic acid, (S)-2-Chloropropionic acid, (S)-α-Chloropropionic acid, 2(S)-Chloropropionic acid, 2-Chloro-L-propionic acid, L-2-Chloropropanoic acid, L-2-Chloropropionic acid, S-(-)-2-Chloropropionic acid, α-L-Chloropropionic acid
InChI Formula InChI=1S/C3H5ClO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m0/s1
SMILES C[C@H](Cl)C(=O)O
IUPAC Name (2S)-2-chloropropanoic acid
Molecular Weight (g/mol) 108.52
Formula Weight 107.9978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.