missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic acid, 98%

Product Code. 10412821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412821 5 g Glass bottle 5g
10637494 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412821 Supplier Thermo Scientific Acros Supplier No. 254970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4°C
Color Yellow
Density 1.2400g/mL
Boiling Point 187°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.436
Quantity 5 g
Specific Gravity 1.24
Specific Rotation -14.5
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 108.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Chloropropionic acid, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-2-Chloropropionic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.