missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Chloromandelic acid, 95%

Product Code. 15457658
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457658 5 g 5g
15467658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457658 Supplier Thermo Scientific Alfa Aesar Supplier No. H61152.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52950-19-3 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52950-19-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798437
InChI Key RWOLDZZTBNYTMS-ZETCQYMHSA-N
Synonym s-+-2-chloromandelic acid, s-2-2-chlorophenyl-2-hydroxyacetic acid, s-2-chloromandelic acid, s-o-chloromandelicacid, 2-chloro-l-mandelic acid, 2s-2-2-chlorophenyl-2-hydroxy-acetic acid, s-2-chlorophenyl hydroxy acetic acid, pubchem5743, s-o-chloromandelic acid, ksc914s2j
PubChem CID 6922874
IUPAC Name (2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl

Specifications

Melting Point 118°C to 122°C
Color Brown to White
Quantity 5 g
Physical Form Powder
Formula Weight 186.59
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-Chloromandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.