missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Butanol, 98+%

Product Code. 11309025
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309025 5 g 5g
11399015 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309025 Supplier Thermo Scientific Alfa Aesar Supplier No. L14164.06
  • ONLINE EXCLUSIVE PRICE
    £237.66
    £195.65 / 5g
    Save £42.01
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4221-99-2 | C4H10O | 74.123 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064281
InChI Key BTANRVKWQNVYAZ-BYPYZUCNSA-N
Synonym s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
PubChem CID 444683
ChEBI CHEBI:45475
IUPAC Name (2S)-butan-2-ol
SMILES CCC(C)O

Specifications

Density 0.806
Quantity 5 g
Flash Point 26°C (79°F)
Odor Alcohol-like
Refractive Index 1.398
UN Number UN1120
Merck Index 141541
Boiling Point 99°C to 100°C
Formula Weight 74.12
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.