missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate, Thermo Scientific™

Product Code. p-7028536
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445660 100 mg 100mg
11455660 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445660 Supplier Thermo Scientific Alfa Aesar Supplier No. H25907.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301706-56-8
Molecular Formula C19H34N2O8
Molecular Weight (g/mol) 418.487
MDL Number MFCD09953464
InChI Key HCNKZGJFFYKTGU-GYDOPSIJSA-N
Synonym 2s-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2s-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2s-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate
PubChem CID 44119501
IUPAC Name (E)-but-2-enedioic acid;tert-butyl N-[[(2S)-4,4-diethoxypiperidin-2-yl]methyl]carbamate
SMILES CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O

Specifications

Quantity 100 mg
Formula Weight 418.49
Chemical Name or Material (S)-(-)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.