missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Benzylsuccinic acid 1-methyl ester, 98+%, ee 98+%, Thermo Scientific Chemicals

Product Code. 11354945
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11354945

Brand: Thermo Scientific Alfa Aesar L18265.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182247-45-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01311761
InChI Key BUNMUVFKMIOEQU-JTQLQIEISA-N
Synonym s-3-benzyl-4-methoxy-4-oxobutanoic acid, s---2-benzylsuccinic acid 1-methyl ester, 3s-3-benzyl-4-methoxy-4-oxobutanoic acid, s-2-benzylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-4-phenylbutanoic acid, butanedioic acid, phenylmethyl-, 1-methyl ester, 2s-9ci
PubChem CID 7006556
IUPAC Name (3S)-3-benzyl-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(CC1=CC=CC=C1)CC(=O)O

Specifications

Quantity 250 mg
Beilstein 7638790
Formula Weight 222.24
Percent Purity ≥98%
Assay ee ≥98%
Chemical Name or Material (S)-(-)-2-Benzylsuccinic acid 1-methyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.