missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Azetidinecarboxylic acid, 99+%

Product Code. 10192220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
250mg
50mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192220 100 mg Glass bottle 100mg
10072041 50 mg Glass bottle 50mg
10624092 250 mg Glass bottle 250mg
10731735 1 g Glass bottle 1g
11986591 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10192220 Supplier Thermo Scientific Acros Supplier No. 105141000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2133-34-8 | C4H7NO2 | 101.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-34-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
InChI Key IADUEWIQBXOCDZ-VKHMYHEASA-N
Synonym l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
PubChem CID 16486
ChEBI CHEBI:6198
IUPAC Name (2S)-azetidine-2-carboxylic acid
SMILES C1CNC1C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 206°C to 207°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 mg
Merck Index 10,911
Specific Rotation Condition −125° (25°C c=1,water)
Specific Rotation -125
Solubility Information Solubility in water: 5% in water
Formula Weight 101.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.