missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminooctane, ChiPros™ 99+%, ee 99+%

Product Code. p-7058505
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11364216

Brand: Thermo Scientific Alfa Aesar L19309.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34566-04-6 | C8H19N | 129.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34566-04-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD01091021
InChI Key HBXNJMZWGSCKPW-QMMMGPOBSA-N
Synonym s-2-aminooctane, s-2-octanamine, s-2-octylamine, 2-octanamine, 2s, unii-7vj4uq98e4, 2s-octan-2-amine, s-+-2-aminooctane, +-2-octylamine, +-2-aminooctane, 2-octanamine, s
PubChem CID 6999019
IUPAC Name (2S)-octan-2-amine
SMILES CCCCCCC(C)N

Specifications

Melting Point <-20°C
Boiling Point 163°C to 165°C
Flash Point 50°C (122°F)
Quantity 1 g
UN Number UN2734
Beilstein 1719319
Formula Weight 129.25
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminooctane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.