missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminooctane, ChiPros™ 99+%, ee 99+%

Product Code. 11384216
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384216 25 g 25g
11364216 1 g 1g
11374216 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384216 Supplier Thermo Scientific Alfa Aesar Supplier No. L19309.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 34566-04-6 | C8H19N | 129.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34566-04-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD01091021
InChI Key HBXNJMZWGSCKPW-QMMMGPOBSA-N
Synonym s-2-aminooctane, s-2-octanamine, s-2-octylamine, 2-octanamine, 2s, unii-7vj4uq98e4, 2s-octan-2-amine, s-+-2-aminooctane, +-2-octylamine, +-2-aminooctane, 2-octanamine, s
PubChem CID 6999019
IUPAC Name (2S)-octan-2-amine
SMILES CCCCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.77
Boiling Point 163°C to 165°C
Flash Point 50°C (122°F)
Refractive Index 1.4235
Quantity 25 g
UN Number UN2734
Beilstein 1719319
Sensitivity Air Sensitive
Formula Weight 129.25
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.