missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine, 95%

Product Code. 15483678
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483678 5 g 5g
15473678 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483678 Supplier Thermo Scientific Alfa Aesar Supplier No. H57137.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22795-99-9 | C7H16N2 | 128.219 g/mol

It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Hygroscopic. Store away from acids, water/moisture, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22795-99-9
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00191371
InChI Key UNRBEYYLYRXYCG-ZETCQYMHSA-N
Synonym s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590
PubChem CID 643457
IUPAC Name [(2S)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

Density 0.919
Boiling Point 50°C to 54°C
Flash Point 57°C (135°F)
Refractive Index 1.466
Quantity 5 g
UN Number UN2733
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform,methanol. Slightly soluble in water.
Optical Rotation −95° (c=1 in Methanol)
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.