missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminomethyl-1-Boc-pyrrolidine, 97%

Product Code. 15499457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499457 1 g 1g
15409467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499457 Supplier Thermo Scientific Alfa Aesar Supplier No. H52727.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119020-01-8 | C10H20N2O2 | 200.28 g/mol

(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.

Solubility
Miscible with N-methylpyrrolidinone.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119020-01-8
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03419257
InChI Key SOGXYCNKQQJEED-QMMMGPOBSA-N
Synonym s-1-boc-2-aminomethyl pyrrolidine, s-1-n-boc-2-aminomethyl pyrrolidine, s-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-aminomethyl pyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyrrolidine, s-2-aminomethyl-1-boc-pyrrolidine, tert-butyl s-2-aminomethylpyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512533
IUPAC Name tert-butyl (2S)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1CN

Specifications

Boiling Point 98°C to 112°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Solubility Information Miscible with N-methylpyrrolidinone.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-2-Aminomethyl-1-Boc-pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.