missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminobutyramide Hydrochloride, TRC

Product Code. 30133086
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133086 25 g 25g
30137219 5 g 5g
17933985 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133086 Supplier Toronto Research Chemicals Supplier No. TRCA60289025G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7682-20-4
Molecular Formula C4 H10 N2 O . Cl H
Molecular Weight (g/mol) 138.6
Synonym Butanamide, 2-amino-, hydrochloride (1:1), (2S)-, Butanamide, 2-amino-, monohydrochloride, (2S)- (9CI), Butanamide, 2-amino-, monohydrochloride, (S)-, Butyramide, 2-amino-, monohydrochloride, L- (8CI), (S)-(+)-2-Aminobutanamide hydrochloride, (S)-2-Aminobutanamide hydrochloride, (S)-2-Aminobutyramide hydrochloride, Levetiracetam Related Compound B, Levetiracetam Imp. G (EP)
IUPAC Name (2S)-2-aminobutanamide;hydrochloride
SMILES Cl.CC[C@H](N)C(=O)N

Specifications

InChI Formula InChI=1S/C4H10N2O.ClH/c1-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H2,6,7);1H/t3-;/m0./s1
Quantity 25 g
Formula Weight 138.056
Shipping Condition Room Temperature
Chemical Name or Material (S)-2-Aminobutanamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.