missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminobutanoic Acid, Mikromol™

Product Code. 30049922
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049922 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049922 Supplier Mikromol Supplier No. MM1372.070100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(S)-2-Aminobutanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Aminobutyric Acid
CAS 1492-24-6
Analyte or Component Names L-Aminobutyric Acid
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O2
Certifications/Compliance ISO 9001
Synonym Butanoic acid, 2-amino-, (2S)-, Butanoic acid, 2-amino-, (S)-, Butyric acid, 2-amino-, L- (8CI), (2S)-2-Aminobutanoic acid, (+)-2-Aminobutanoic acid, (+)-2-Aminobutyric acid, (+)-α-Aminobutyric acid, (S)-(+)-α-Aminobutyric acid, (S)-2-Aminobutanoic acid, (S)-2-Aminobutyric acid, L-2-Amino-n-butyric acid, L-2-Aminobutanoic acid, L-2-Aminobutyric acid, L-Butyrine, L-Ethylglycine, L-Homoalanine, L-α-Amino-n-butyric acid, L-α-Aminobutyric acid, NSC 97060, S-Butyrine
InChI Formula InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1
SMILES CC[C@H](N)C(=O)O
IUPAC Name (2S)-2-aminobutanoic acid
Molecular Weight (g/mol) 103.12
Quantity 100 mg
Formula Weight 103.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.