missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutane, 98%

Product Code. 11392417
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392417 250 mg 250mg
11372417 1 g 1g
11382417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392417 Supplier Thermo Scientific Alfa Aesar Supplier No. L10069.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-49-5 | C4H11N | 73.139 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-49-5
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00064417
InChI Key BHRZNVHARXXAHW-BYPYZUCNSA-N
Synonym s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s
PubChem CID 6713753
IUPAC Name (2S)-butan-2-amine
SMILES CCC(C)N

Specifications

Density 0.722
pH 12.5
Boiling Point 63°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 250 mg
UN Number UN2733
Beilstein 1718760
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Fully miscible in water.
Optical Rotation +7.5° (Neat)
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Aminobutane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-2-Aminobutane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (S)-sec-Butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.