missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals

Product Code. 15426189
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 15426189

Brand: Thermo Scientific Alfa Aesar J64075.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A NMDA agonist

A NMDA agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23052-81-5
Molecular Formula C4H10NO5P
Molecular Weight (g/mol) 183.1
MDL Number MFCD00083244
InChI Key DDOQBQRIEWHWBT-VKHMYHEASA-N
Synonym l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid
PubChem CID 179394
IUPAC Name (2S)-2-amino-4-phosphonobutanoic acid
SMILES C(CP(=O)(O)O)C(C(=O)O)N

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Solubility Information Soluble to 5 mM in water and to 100 mM in 1eq. NaOH
Optical Rotation +24° (c=1 in 6N HCl)
Formula Weight 183.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (S)-(+)-2-Amino-4-phosphonobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.