missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-4-pentenoic acid hydrochloride, Thermo Scientific™

Product Code. 10109869
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10109869 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10109869 Supplier Thermo Scientific Maybridge Supplier No. MAY00253DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 195316-72-4
Packaging Amber Glass Bottle
Molecular Formula C5H10ClNO2
Quantity 1 g
Synonym s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s
InChI Key DIDZZOWASZMNQW-WCCKRBBISA-N
SMILES C=CCC(C(=O)O)N.Cl
IUPAC Name (2S)-2-aminopent-4-enoic acid;hydrochloride
Molecular Weight (g/mol) 151.59
PubChem CID 44630067
Formula Weight 151.59
Percent Purity 97%
Chemical Name or Material (S)-(-)-2-Amino-4-pentenoic acid hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.