missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid, 98%, 94% ee

Product Code. 10379060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10379060 250 mg Glass bottle 250mg
11945421 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10379060 Supplier Thermo Scientific Acros Supplier No. 299142500
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 2280-27-5 | C5H11NO3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2280-27-5
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
InChI Key LDRFQSZFVGJGGP-GSVOUGTGSA-N
Synonym s-2-amino-3-hydroxy-3-methylbutanoic acid, 3-hydroxy-l-valine, s-+-2-amino-3-hydroxy-3-methylbutanoic acid, beta-hydroxy-l-valine, valine, 3-hydroxy, 2s-2-amino-3-hydroxy-3-methylbutanoic acid, 2s-2-amino-3-hydroxy-3-methyl-butanoic acid, 3-hydroxyvaline, 3-methylthreonine, 3-hydroxy-l-val
PubChem CID 192763
IUPAC Name (2S)-2-amino-3-hydroxy-3-methylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)O

Specifications

Color White
Enantiomeric Excess 93% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3C(CH3)(OH)CH(NH2)COOH
Quantity 250 mg
Specific Rotation Condition − 11.75 (25.00°C c=2,6N HCl)
Specific Rotation − 11.75
Formula Weight 133.15
Percent Purity 98%,94% ee
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.