missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 15499715
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499715 5 g 5g
15409725 250 mg 250mg
15489715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499715 Supplier Thermo Scientific Alfa Aesar Supplier No. H52297.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61366-43-6
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
MDL Number MFCD07368914
InChI Key BJWHQBTUHVIRJJ-UHFFFAOYNA-N
Synonym s-2-amino-3-benzyloxy propan-1-ol hydrochloride, s-2-amino-3-benzyloxy-1-propanol hydrochloride, 2s-2-amino-3-benzyloxy propan-1-ol hydrochloride, s---2-amino-3-benzyloxy-1-propanol??hcl, s-2-amino-3-benzyloxy-propan-1-ol hydrochloride, 2s-2-amino-3-benzyloxy propan-1-ol-hydrogen chloride 1/1
PubChem CID 53338757
SMILES Cl.NC(CO)COCC1=CC=CC=C1

Specifications

Quantity 5 g
IUPAC Name (2S)-2-amino-3-phenylmethoxypropan-1-ol;hydrochloride
Formula Weight 217.7
Percent Purity ≥98%
Chemical Name or Material (S)-2-Amino-3-benzyloxy-1-propanol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.