missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+%

Product Code. 15468937
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468937 5 g 5g
15478937 250 mg 250mg
15458937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468937 Supplier Thermo Scientific Alfa Aesar Supplier No. H52304.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58577-88-1 | C10H15NO2 | 181.235 g/mol

(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58577-88-1
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD04112477
InChI Key ZJUOMDNENVWMPL-JTQLQIEISA-N
Synonym s-2-amino-3-benzyloxy-1-propanol, h-d-ser bzl-ol, 2s-2-amino-3-benzyloxy propan-1-ol, s-2-amino-3-benzyloxy propan-1-ol, 1-propanol, 2-amino-3-phenylmethoxy-, 2s, s---2-amino-3-benzyloxy-1-propanol, 98+%, ambotzhal1035, s---2-amino-3-benzyloxy-1-propal, s---2-amino-3-benzyloxy-1-propanol
PubChem CID 11401131
IUPAC Name (2S)-2-amino-3-phenylmethoxypropan-1-ol
SMILES C1=CC=C(C=C1)COCC(CO)N

Specifications

Quantity 5 g
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 181.23
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Amino-3-benzyloxy-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.