missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic Acid (5-Hydroxytryptophan), Mikromol™
Description
(S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic Acid (5-Hydroxytryptophan)
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-L-tryptophan |
| CAS | 9-8-4350 |
| Analyte or Component Names | 5-Hydroxy-L-tryptophan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H12 N2 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | (S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic Acid, N-Acetyltryptophan Imp. D (EP), Tryptophan Imp. D (EP), 5-Hydroxytryptophan, Tryptophan, 5-hydroxy-, L- (8CI), (S)-5-Hydroxytryptophan, 5-Hydroxy-L-tryptophan, 5-Hydroxyl-L-tryptophan, Cincofarm, L-5-HTP, L-5-Hydroxytryptophan, Levothym, Levotinine, Oxitriptan, Oxyfan, Pretonine, Quietim, Serotonyl, Telesol, Tript-Oh, Triptene, N-Acetyltryptophan Impurity D, Tryptophan Impurity D |
| InChI Formula | InChI=1S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14H,3,12H2,(H,15,16)/t9-/m0/s1 |
| SMILES | N[C@@H](Cc1c[nH]c2ccc(O)cc12)C(=O)O |
| IUPAC Name | (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?