missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-2-methyl-4-pentenoic acid, 98%, ee 99+%

Product Code. 15491766
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15491766

Brand: Thermo Scientific Alfa Aesar H54922.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96886-55-4 | C6H11NO2 | 129.159 g/mol

It is used in organic synthesis and as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis and as an pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96886-55-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00145248
InChI Key QMBTZYHBJFPEJB-LURJTMIESA-N
Synonym s-2-amino-2-methylpent-4-enoic acid, s-2-amino-2-methyl-4-pentenoic acid, l-alpha-allylalanine, 2s-2-amino-2-methylpent-4-enoic acid, alpha-allyl-l-ala, r-2-amino-2-methyl-4-pentenoic acid, alpha-methyl-l-allylglycine, s---alpha-allylalanine, 4-pentenoic acid, 2-amino-2-methyl-, 2s, s---2-amino-2-methyl-4-pentenoic acid
PubChem CID 2724753
IUPAC Name (2S)-2-amino-2-methylpent-4-enoic acid
SMILES CC(CC=C)(C(=O)O)N

Specifications

Melting Point 285°C
Quantity 1 g
Solubility Information Very soluble in water.
Optical Rotation −14° (c=1.3 in 1N HCl)
Formula Weight 129.16
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-2-Amino-2-methyl-4-pentenoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.