missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-propanol, 98%

Product Code. 10325880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10325880 10 g Glass bottle 10g
10315880 1 g Glass bottle 1g
10445365 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10325880 Supplier Thermo Scientific Acros Supplier No. 104450100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2749-11-3 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2749-11-3
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064412
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
PubChem CID 80307
ChEBI CHEBI:78502
IUPAC Name (2S)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9650g/mL
Boiling Point 173°C to 176°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(NH2)CH2OH
Refractive Index 1.4480 to 1.451
Quantity 10 g
Beilstein 04, I, 432
Merck Index 10, 472
Specific Gravity 0.965
Specific Rotation + 22
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 75.11
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material (S)-(+)-2-Amino-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-2-Amino-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.