missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-propanol, 98%

Product Code. 11421407
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421407 25 g 25g
11401407 1 g 1g
11411407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421407 Supplier Thermo Scientific Alfa Aesar Supplier No. B24916.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2749-11-3 | C3H10NO | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2749-11-3
MDL Number MFCD00064412

Specifications

Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Molecular Formula C5H13Br2N
Refractive Index 1.4498
Quantity 25 g
UN Number UN2735
Beilstein 1718865
Sensitivity Air sensitive; Hygroscopic
Synonym 2-bromoethyl trimethylammonium bromide, 2-bromo-n,n,n-trimethylethanaminium bromide, 2-bromoethyltrimethylammonium bromide, bromcholin, hypercyl, kathesin, btab, btab the plant regulator, bcb the plant regulator, unii-kh1i7ru18l
InChI Key OINMNSFDYTYXEQ-UHFFFAOYSA-M
Optical Rotation +22°C (c = 2 in Ethanol)
SMILES C[N+](C)(C)CCBr.[Br-]
IUPAC Name 2-bromoethyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 246.97
PubChem CID 17689
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Amino-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.