missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Acetoxysuccinic anhydride, 98%, Thermo Scientific™

Product Code. 11478530
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478530 10 g 10g
11488530 2 g 2g
11498530 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478530 Supplier Thermo Scientific Alfa Aesar Supplier No. 044366.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59025-03-5
Molecular Formula C6H6O5
Molecular Weight (g/mol) 158.109
MDL Number MFCD00082708
InChI Key SSWJHSASZZAIAU-BYPYZUCNSA-N
Synonym s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate
PubChem CID 11126528
IUPAC Name [(3S)-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1CC(=O)OC1=O

Specifications

Melting Point 56°C to 58°C
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Reacts in water.
Formula Weight 158.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)- (-)-2-Acetoxysuccinic anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.