missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%

Product Code. 11302307
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11302307

Brand: Thermo Scientific Alfa Aesar L09855.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22204-53-1 | C14H14O3 | 230.263 g/mol

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22204-53-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD00010500
InChI Key CMWTZPSULFXXJA-VIFPVBQESA-N
Synonym naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno
PubChem CID 156391
ChEBI CHEBI:7476
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O

Specifications

Melting Point 154°C to 156°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 3591067
Merck Index 14,6417
Solubility Information Insoluble in water.
Optical Rotation +66° (c=1 in Chloroform)
Formula Weight 230.26
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Naproxen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.