missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%

Product Code. 10330804 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330804 1 mL Glass bottle 1mL
10774154 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10330804 Supplier Thermo Scientific Acros Supplier No. 389960010
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

(S)-2, 5-Dihydro-3, 6-dimethoxy-2-isopropylpyrazine, 97+%, C9H16N2O2, CAS Number-78342-42-4 | CAS: 78342-42-4 | C9H16N2O2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78342-42-4
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00066229
InChI Key FCFWEOGTZZPCTO-QMMMGPOBSA-N
Synonym s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, 2s-+-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, 2s-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2s, 2s-2,5-dihydro-2-isopropyl-3,6-dimethoxypyrazine, 2s-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, 3s-3-isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, s-2,5-dihydro-3,6-dimethoxy-2-iso-propylpyrazine, 2s-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
PubChem CID 736065
IUPAC Name (2S)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
SMILES CC(C)C1C(=NCC(=N1)OC)OC

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0280g/mL
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Quantity 1 mL
Specific Gravity 1.028
Specific Rotation +96° to +107° (20°C, 589nm) (C=1, EtOH)
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 184.24
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material (S)-2, 5-Dihydro-3, 6-dimethoxy-2-isopropylpyrazine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.