missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(4-Chlorophenyl)-3-methylbutanoic Acid, TRC

Product Code. 30135342
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135342 10 mg 10mg
30135376 50 mg 50mg
30006605 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135342 Supplier Toronto Research Chemicals Supplier No. TRCC38571310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55332-38-2
Molecular Formula C11 H13 Cl O2
Molecular Weight (g/mol) 212.67
Synonym Benzeneacetic acid, 4-chloro-α-(1-methylethyl)-, (S)-, (αS)-4-Chloro-α-(1-methylethyl)benzeneacetic acid, (+)-(S)-2-(4-Chlorophenyl)-3-methylbutanoic acid, (+)-2-(4-Chlorophenyl)-3-methylbutyric acid, (+)-2-(4-Chlorophenyl)isovaleric acid, (+)-α-(p-Chlorophenyl)isovaleric acid, (2S)-2-(4-Chlorophenyl)-3-methylbutanoic acid, (S)-2-(4-Chlorophenyl)-3-methylbutyric acid, (S)-2-(4-Chlorophenyl)isovaleric acid, (S)-Fenvaleric acid, 2-S-(+)-(4-Chlorophenyl)-3-methylbutanoic Acid
IUPAC Name (2S)-2-(4-chlorophenyl)-3-methylbutanoic acid
SMILES CC(C)[C@H](C(=O)O)c1ccc(Cl)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H13ClO2/c1-7(2)10(11(13)14)8-3-5-9(12)6-4-8/h3-7,10H,1-2H3,(H,13,14)/t10-/m0/s1
Formula Weight 212.0604
Chemical Name or Material Esfenvalerate free acid metabolite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.