missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid, TRC

Product Code. 30357393
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357393 500 mg 500mg
30349038 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357393 Supplier Toronto Research Chemicals Supplier No. TRCC596875500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid
CAS 76496-63-4
Molecular Formula C8H7O3Cl
Quantity 500 mg
InChI Formula InChI=1S/C8H7ClO3/c9-6-3-1-5(2-4-6)7(10)8(11)12/h1-4,7,10H,(H,11,12)/t7-/m0/s1
SMILES O[C@H](C(=O)O)c1ccc(Cl)cc1
IUPAC Name (2S)-2-(4-chlorophenyl)-2-hydroxyacetic acid
Molecular Weight (g/mol) 186.59
Formula Weight 186.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.