missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,3-O-Isopropylideneglycerol, 98%

Product Code. 11425957
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425957 10 g 10g
11415957 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425957 Supplier Thermo Scientific Alfa Aesar Supplier No. B24488.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22323-82-6 | C6H12O3 | 132.16 g/mol

(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2,3-O-Isopropylideneglycerol acts as a MEK inhibitor with antitumor activity. It is also employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22323-82-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00063239
InChI Key RNVYQYLELCKWAN-UHFFFAOYNA-N
Synonym s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol
PubChem CID 736057
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(C)OCC(CO)O1

Specifications

Density 1.066
Boiling Point 82°C to 83°C (14 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.435
Quantity 10 g
Beilstein 80118
Merck Index 14,5213
Solubility Information Miscible with water and chloroform.
Optical Rotation +11.5° (c=5 in Methanol)
Formula Weight 132.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2,3-O-Isopropylideneglycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.