missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,3-Diaminopropionic acid monohydrochloride, 95%

Product Code. 15487066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487066 1 g 1g
15497066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487066 Supplier Thermo Scientific Alfa Aesar Supplier No. H62128.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1482-97-9 | C3H9ClN2O2 | 140.567 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1482-97-9
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00065497
InChI Key SKWCZPYWFRTSDD-DKWTVANSSA-N
Synonym 3-amino-l-alanine hydrochloride, s-2,3-diaminopropanoic acid hydrochloride, h-dap-oh.hcl, l-2,3-diaminopropionic acid hydrochloride, l +-2,3-diaminopropionic acid hydrochloride, l-3-aminoalanine hydrochloride, h-dapa-oh.hcl, s-+-2,3-diaminopropionic acid hydrochloride, l-alanine, 3-amino-, monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +
PubChem CID 12495315
IUPAC Name (2S)-2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 231°C to 233°C (decomposition)
Color White
Quantity 1 g
Beilstein 4263944
Sensitivity Hygroscopic
Formula Weight 140.57
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material (S)-2,3-Diaminopropionic acid monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.