missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%

Product Code. 10145460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10145460 1 g Glass bottle 1g
10026043 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10145460 Supplier Thermo Scientific Acros Supplier No. 296470010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17553-86-5 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17553-86-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00012272
InChI Key FNYAZSZTENLTRT-JTQLQIEISA-N
Synonym 7as-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione, s-+-2,3,7,7a-tetrahydro-7a-methyl-1h-indene-1,5 6h-dione, 1h-indene-1,5 6h-dione, 2,3,7,7a-tetrahydro-7a-methyl-, 7as, hajos dione, s-hajos dione, s-hajos ketone, s-hajos-wiechert ketone, +-hajos-parrish ketone, +-hajos-parrish diketone, 7as-7a-methyl-2,3,7,7a-tetrahydro-1h-indene-1,5 6h-dione
PubChem CID 736943
IUPAC Name (7aS)-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
SMILES CC12CCC(=O)C=C1CCC2=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 55.0°C to 64.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +331° (20°C c=1,toluene)
Specific Rotation + 331.00
Formula Weight 164.2
Percent Purity 90%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material (S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.